Anti-Haptoglobin Mouse Monoclonal Antibody [Clone: HP/3835]

Lieferant: ANTIBODIES.COM
A278234-100
ANTIA278234-100EA 520 EUR
ANTIA278234-100
Anti-Haptoglobin Mouse Monoclonal Antibody [Clone: HP/3835]
Antikörper
Mouse monoclonal [HP/3835] antibody to Haptoglobin for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Haptoglobin
Clonality: Monoclonal
Clone: HP/3835
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR