Anti-Haptoglobin Mouse Monoclonal Antibody [clone: HP/3839]
Lieferant: ANTIBODIES.COM
A278236-100
ANTIA278236-100EA 520 EUR
ANTIA278236-100
Anti-Haptoglobin Mouse Monoclonal Antibody [clone: HP/3839]
Antikörper
Mouse monoclonal [HP/3839] antibody to Haptoglobin for IHC-P with samples derived from human.
  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Haptoglobin
Clonality: Monoclonal
Clone: HP/3839
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human