Anti-IL-1 beta Rabbit Recombinant Antibody [clone: IL1B/7049R]

Lieferant: ANTIBODIES.COM
A278592-100
ANTIA278592-100EA 520 EUR
ANTIA278592-100
Anti-IL-1 beta Rabbit Recombinant Antibody [clone: IL1B/7049R]
Antikörper
Recombinant rabbit monoclonal [IL1B/7049R] antibody to IL-1 beta for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: IL-1 beta
Clonality: Recombinant
Clone: IL1B/7049R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR