Anti-RRM1 Rabbit Recombinant Antibody [clone: RRM1/4372R]

Lieferant: ANTIBODIES.COM
A278647-100
ANTIA278647-100EA 520 EUR
ANTIA278647-100
Anti-RRM1 Rabbit Recombinant Antibody [clone: RRM1/4372R]
Antikörper
Recombinant rabbit monoclonal [RRM1/4372R] antibody to RRM1 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: RRM1
Clonality: Recombinant
Clone: RRM1/4372R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR