Anti-CD172 gamma Mouse Monoclonal Antibody [clone: OX-119] (PE (Phycoerythrin))

Lieferant: ANTIBODIES.COM
A285822-100
ANTIA285822-100EA 435 EUR
ANTIA285822-100
Anti-CD172 gamma Mouse Monoclonal Antibody [clone: OX-119] (PE (Phycoerythrin))
Antikörper
Mouse monoclonal [OX-119] antibody to CD172 gamma (PE) for Flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: The reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD172 gamma
Clonality: Monoclonal
Clone: OX-119
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR