Anti-HLA C Mouse Monoclonal Antibody [Clone: DT-9]
Lieferant: ANTIBODIES.COM
A285864-100
ANTIA285864-100EA 340 EUR
ANTIA285864-100
Anti-HLA C Mouse Monoclonal Antibody [Clone: DT-9]
Antikörper
Mouse monoclonal [DT-9] antibody to HLA C for IP and Flow cytometry with samples derived from non-human primates and human.
  • Validated applications: IP, Flow cytometry
  • Recommended dilutions: Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: HLA C
Clonality: Monoclonal
Clone: DT-9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Non-Human Primates, Human