Anti-Rabbit IgG H&L Goat Polyclonal Antibody (Cyanine 5)

Lieferant: Antibodies.com
A294878-500
ANTIA294878-500EA 240 EUR
ANTIA294878-500
Anti-Rabbit IgG H&L Goat Polyclonal Antibody (Cyanine 5)
Antikörper
Goat anti-rabbit IgG H&L (Cyanine 5) secondary antibody for WB, IHC, ICC, Flow cytometry and IF with samples derived from rabbit.

  • Validated applications: WB, IHC, ICC, Flow cytometry, IF
  • Recommended dilutions: WB: 1:1000 to 1:15000, IHC: 1:50 to 1:500, ICC: 1:50 to 1:500, Flow cytometry: 1:50 to 1:200, IF: 1:50 to 1:500

Type: Secondary
Antigen: Rabbit IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR