Anti-Human IgG H&L Goat Polyclonal Antibody (Cyanine 5)
Lieferant: ANTIBODIES.COM
A301237-500
ANTIA301237-500EA 240 EUR
ANTIA301237-500
Anti-Human IgG H&L Goat Polyclonal Antibody (Cyanine 5)
Antikörper
Goat anti-human IgG H&L (Cyanine 5) secondary antibody for IHC, ICC, Flow cytometry and IF with samples derived from human.
  • Validated applications: IHC, ICC, Flow cytometry, IF
  • Recommended dilutions: Flow cytometry: 1:50 to 1:200, ICC: 1:50 to 1:500, IF: 1:50 to 1:500, IHC: 1:50 to 1:500
Type: Secondary
Antigen: Human IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human