Anti-4-Hydroxy-2-hexenal Mouse Monoclonal Antibody [Clone: 5C11.1]

Lieferant: ANTIBODIES.COM
A304706-100
ANTIA304706-100EA 580 EUR
ANTIA304706-100
Anti-4-Hydroxy-2-hexenal Mouse Monoclonal Antibody [Clone: 5C11.1]
Antikörper
Mouse monoclonal [5C11.1] antibody to 4-Hydroxy-2-hexenal for WB and ELISA with samples derived from species independent.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:1000, ELISA: 1:1000

Type: Primary
Antigen: 4-Hydroxy-2-hexenal
Clonality: Monoclonal
Clone: 5C11.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Species Independent
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR