Anti-4-Hydroxy-2-hexenal Mouse Monoclonal Antibody [clone: 6F10]
Lieferant: ANTIBODIES.COM
A304729-100
ANTIA304729-100EA 580 EUR
ANTIA304729-100
Anti-4-Hydroxy-2-hexenal Mouse Monoclonal Antibody [clone: 6F10]
Antikörper
Mouse monoclonal [6F10] antibody to 4-Hydroxy-2-hexenal for WB, ICC/IF, ELISA, FACS and Flow cytometry with samples derived from species independent.
  • Validated applications: WB, ICC/IF, ELISA, FACS, Flow cytometry
  • Recommended dilutions: WB: 1:1000, ICC/IF: 1:50, ELISA: 1:1000, FACS: 1:50, FCM: 1:50
Type: Primary
Antigen: 4-Hydroxy-2-hexenal
Clonality: Monoclonal
Clone: 6F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Species Independent