Anti-Crotonaldehyde Mouse Monoclonal Antibody [Clone: 2A8.1]
Lieferant: ANTIBODIES.COM
A304730-100
ANTIA304730-100EA 580 EUR
ANTIA304730-100
Anti-Crotonaldehyde Mouse Monoclonal Antibody [Clone: 2A8.1]
Antikörper
Mouse monoclonal [2A8.1] antibody to Crotonaldehyde for WB and ELISA with samples derived from species independent.
  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:1000, ELISA: 1:1000
Type: Primary
Antigen: Crotonaldehyde
Clonality: Monoclonal
Clone: 2A8.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Species Independent