Anti-Crotonaldehyde Mouse Monoclonal Antibody [clone: 2D3.1]

Lieferant: ANTIBODIES.COM
A304731-100
ANTIA304731-100EA 580 EUR
ANTIA304731-100
Anti-Crotonaldehyde Mouse Monoclonal Antibody [clone: 2D3.1]
Antikörper
Mouse monoclonal [2D3.1] antibody to Crotonaldehyde for WB and ELISA with samples derived from species independent.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:1000, ELISA: 1:1000

Type: Primary
Antigen: Crotonaldehyde
Clonality: Monoclonal
Clone: 2D3.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Species Independent
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR