Anti-Acrolein Mouse Monoclonal Antibody [Clone: 2H2]
Lieferant: ANTIBODIES.COM
A304754-100
ANTIA304754-100EA 580 EUR
ANTIA304754-100
Anti-Acrolein Mouse Monoclonal Antibody [Clone: 2H2]
Antikörper
Mouse monoclonal [2H2] antibody to Acrolein for WB, ICC/IF, FACS, Flow cytometry and ELISA with samples derived from species independent.
  • Validated applications: WB, ICC/IF, FACS, Flow cytometry, ELISA
  • Recommended dilutions: WB: 1:1000, ICC/IF: 1:50, ELISA: 1:1000
Type: Primary
Antigen: Acrolein
Clonality: Monoclonal
Clone: 2H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Species Independent