Anti-LC3A Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A304958-100
ANTIA304958-100EA 475 EUR
ANTIA304958-100
Anti-LC3A Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to LC3A for WB and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: WB: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: LC3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR