Anti-Aquaporin 2 Rabbit Polyclonal Antibody
Lieferant: ANTIBODIES.COM
A305443-100
ANTIA305443-100EA 415 EUR
ANTIA305443-100
Anti-Aquaporin 2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to Aquaporin 2 for IHC with samples derived from mouse.
  • Validated applications: IHC
  • Recommended dilutions: IHC: 1:50 to 1:200
Type: Primary
Antigen: Aquaporin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse