Anti-PHGDH/Malate dehydrogenase Rabbit Polyclonal Antibody

Lieferant: Antibodies.com
A305698-100
ANTIA305698-100EA 415 EUR
ANTIA305698-100
Anti-PHGDH/Malate dehydrogenase Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to PHGDH/Malate dehydrogenase for WB and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:2000, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: PHGDH/Malate dehydrogenase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR