Anti-U2AF65 Rabbit Monoclonal Antibody [clone: ARC1029]

Lieferant: ANTIBODIES.COM
A305827-100
ANTIA305827-100EA 525 EUR
ANTIA305827-100
Anti-U2AF65 Rabbit Monoclonal Antibody [clone: ARC1029]
Antikörper
Rabbit monoclonal [ARC1029] antibody to U2AF65 for WB, IHC and IP with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, IP
  • Recommended dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, IP: 1:500 to 1:1000

Type: Primary
Antigen: U2AF65
Clonality: Monoclonal
Clone: ARC1029
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR