Anti-SARS-CoV-2 ORF3a Rabbit Polyclonal Antibody
Lieferant: ANTIBODIES.COM
A306060-100
ANTIA306060-100EA 415 EUR
ANTIA306060-100
Anti-SARS-CoV-2 ORF3a Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to SARS-CoV-2 ORF3a for WB with samples derived from SARS-CoV-2.
  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:2000 to 1:6000
Type: Primary
Antigen: SARS-CoV-2 ORF3a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: SARS-CoV-2