Anti-ROX Rabbit Monoclonal Antibody [clone: ARC2336]
Lieferant: ANTIBODIES.COM
A306740-100
ANTIA306740-100EA 525 EUR
ANTIA306740-100
Anti-ROX Rabbit Monoclonal Antibody [clone: ARC2336]
Antikörper
Rabbit monoclonal [ARC2336] antibody to ROX for WB with samples derived from Human, Mouse and Rat.
  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000
Type: Primary
Antigen: ROX
Clonality: Monoclonal
Clone: ARC2336
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat