Anti-GLO1 Rabbit Monoclonal Antibody [clone: ARC0969]

Lieferant: ANTIBODIES.COM
A306878-100
ANTIA306878-100EA 525 EUR
ANTIA306878-100
Anti-GLO1 Rabbit Monoclonal Antibody [clone: ARC0969]
Antikörper
Rabbit monoclonal [ARC0969] antibody to GLO1 for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: GLO1
Clonality: Monoclonal
Clone: ARC0969
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR