Anti-24(S)-Hydroxycholesterol Rabbit Polyclonal Antibody

Lieferant: Antibodies.com
A307250-100
ANTIA307250-100EA 415 EUR
ANTIA307250-100
Anti-24(S)-Hydroxycholesterol Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to 24(S)-Hydroxycholesterol for WB with samples derived from Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:1000

Type: Primary
Antigen: 24(S)-Hydroxycholesterol
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR