Anti-Tec Rabbit Monoclonal Antibody [clone: ARC53949]
Lieferant: ANTIBODIES.COM
A307552-100
ANTIA307552-100EA 525 EUR
ANTIA307552-100
Anti-Tec Rabbit Monoclonal Antibody [clone: ARC53949]
Antikörper
Rabbit monoclonal [ARC53949] antibody to Tec for WB with samples derived from Human, Mouse and Rat.
  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:2000 to 1:10000
Type: Primary
Antigen: Tec
Clonality: Monoclonal
Clone: ARC53949
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat