Anti-LAT (phospho Tyr191) Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A308438-100
ANTIA308438-100EA 535 EUR
ANTIA308438-100
Anti-LAT (phospho Tyr191) Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to LAT (phospho Tyr191) for WB with samples derived from human and mouse.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:1,000

Type: Primary
Antigen: LAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR