Anti-RRM1 Rabbit Monoclonal Antibody [clone: ARC0943]

Lieferant: ANTIBODIES.COM
A309074-100
ANTIA309074-100EA 525 EUR
ANTIA309074-100
Anti-RRM1 Rabbit Monoclonal Antibody [clone: ARC0943]
Antikörper
Rabbit monoclonal [ARC0943] antibody to RRM1 for WB with samples derived from human, mouse and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2,000

Type: Primary
Antigen: RRM1
Clonality: Monoclonal
Clone: ARC0943
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR