Anti-pan crotonyl Lysine Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A309641-100
ANTIA309641-100EA 535 EUR
ANTIA309641-100
Anti-pan crotonyl Lysine Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to pan crotonyl Lysine for WB with samples derived from human, mouse and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:1,000

Type: Primary
Antigen: pan crotonyl Lysine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR