Anti-2'-O-methyluridine Rabbit Monoclonal Antibody [clone: ARC50750]

Lieferant: ANTIBODIES.COM
A310047-100
ANTIA310047-100EA 650 EUR
ANTIA310047-100
Anti-2'-O-methyluridine Rabbit Monoclonal Antibody [clone: ARC50750]
Antikörper
Rabbit monoclonal [ARC50750] antibody to 2'-O-methyluridine for Dot Blot with samples derived from Species Independent.

  • Validated applications: Dot Blot
  • Recommended dilutions: DB: 1:500 to 1:1,000

Type: Primary
Antigen: 2'-O-methyluridine
Clonality: Monoclonal
Clone: ARC50750
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Species Independent
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR