Anti-CD45 Mouse Monoclonal Antibody [clone: PTPRC/6780]

Lieferant: ANTIBODIES.COM
A314705-100
ANTIA314705-100EA 680 EUR
ANTIA314705-100
Anti-CD45 Mouse Monoclonal Antibody [clone: PTPRC/6780]
Antikörper
Mouse monoclonal [PTPRC/6780] antibody to CD45 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: PTPRC/6780
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR