Anti-Haptoglobin Mouse Monoclonal Antibody [clone: HP/4813]
Lieferant: ANTIBODIES.COM
A315052-100
ANTIA315052-100EA 680 EUR
ANTIA315052-100
Anti-Haptoglobin Mouse Monoclonal Antibody [clone: HP/4813]
Antikörper
Mouse monoclonal [HP/4813] antibody to Haptoglobin for IHC-P with samples derived from human.
  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Haptoglobin
Clonality: Monoclonal
Clone: HP/4813
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human