Anti-Haptoglobin Mouse Monoclonal Antibody [clone: HP/3838]
Lieferant: ANTIBODIES.COM
A315057-100
ANTIA315057-100EA 680 EUR
ANTIA315057-100
Anti-Haptoglobin Mouse Monoclonal Antibody [clone: HP/3838]
Antikörper
Mouse monoclonal [HP/3838] antibody to Haptoglobin for WB and IHC-P with samples derived from human.
  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Haptoglobin
Clonality: Monoclonal
Clone: HP/3838
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human