Anti-HLA DR Rabbit Recombinant Antibody [clone: HLA-DRA/6840R]

Lieferant: ANTIBODIES.COM
A315093-100
ANTIA315093-100EA 680 EUR
ANTIA315093-100
Anti-HLA DR Rabbit Recombinant Antibody [clone: HLA-DRA/6840R]
Antikörper
Recombinant rabbit monoclonal [HLA-DRA/6840R] antibody to HLA DR for IHC with samples derived from human.

  • Validated applications: IHC
  • Recommended dilutions: IHC: (Frozen and Formalin to fixed) 1 to 2 µg/ml

Type: Primary
Antigen: HLA DR
Clonality: Recombinant
Clone: HLA-DRA/6840R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR