Anti-NeuN Mouse Recombinant Antibody [clone: rNEUN/8055]

Lieferant: ANTIBODIES.COM
A315389-100
ANTIA315389-100EA 680 EUR
ANTIA315389-100
Anti-NeuN Mouse Recombinant Antibody [clone: rNEUN/8055]
Antikörper
Recombinant mouse monoclonal [rNEUN/8055] antibody to NeuN for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: NeuN
Clonality: Recombinant
Clone: rNEUN/8055
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR