Anti-SPARC Mouse Monoclonal Antibody [clone: OSTN/3756]
Lieferant: ANTIBODIES.COM
A315671-100
ANTIA315671-100EA 680 EUR
ANTIA315671-100
Anti-SPARC Mouse Monoclonal Antibody [clone: OSTN/3756]
Antikörper
Mouse monoclonal [OSTN/3756] antibody to SPARC for IHC-P with samples derived from human.
  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: SPARC
Clonality: Monoclonal
Clone: OSTN/3756
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human