Anti-CD45 Mouse Monoclonal Antibody [clone: PTPRC/6780]
Lieferant: ANTIBODIES.COM
A316089-100
ANTIA316089-100EA 680 EUR
ANTIA316089-100
Anti-CD45 Mouse Monoclonal Antibody [clone: PTPRC/6780]
Antikörper
Mouse monoclonal [PTPRC/6780] antibody to CD45 for IHC-P with samples derived from human.
  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: PTPRC/6780
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human