Anti-Haptoglobin Mouse Monoclonal Antibody [clone: HP/4815]

Lieferant: ANTIBODIES.COM
A316437-100
ANTIA316437-100EA 680 EUR
ANTIA316437-100
Anti-Haptoglobin Mouse Monoclonal Antibody [clone: HP/4815]
Antikörper
Mouse monoclonal [HP/4815] antibody to Haptoglobin for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: Haptoglobin
Clonality: Monoclonal
Clone: HP/4815
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR