Anti-Haptoglobin Mouse Monoclonal Antibody [clone: HP/3834]

Lieferant: ANTIBODIES.COM
A316440-100
ANTIA316440-100EA 680 EUR
ANTIA316440-100
Anti-Haptoglobin Mouse Monoclonal Antibody [clone: HP/3834]
Antikörper
Mouse monoclonal [HP/3834] antibody to Haptoglobin for WB and IHC-P with samples derived from human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: Haptoglobin
Clonality: Monoclonal
Clone: HP/3834
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR