Anti-HLA DR Mouse Recombinant Antibody [clone: rHLA-DRA/8285]

Lieferant: ANTIBODIES.COM
A316474-100
ANTIA316474-100EA 680 EUR
ANTIA316474-100
Anti-HLA DR Mouse Recombinant Antibody [clone: rHLA-DRA/8285]
Antikörper
Recombinant mouse monoclonal [rHLA-DRA/8285] antibody to HLA DR for IHC with samples derived from human.

  • Validated applications: IHC
  • Recommended dilutions: IHC: (Frozen and Formalin - fixed) 1 to 2 µg/ ml

Type: Primary
Antigen: HLA DR
Clonality: Recombinant
Clone: rHLA-DRA/8285
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR