Anti-MASPIN Mouse Monoclonal Antibody [clone: SERPINB5/4978]

Lieferant: ANTIBODIES.COM
A316651-100
ANTIA316651-100EA 680 EUR
ANTIA316651-100
Anti-MASPIN Mouse Monoclonal Antibody [clone: SERPINB5/4978]
Antikörper
Mouse monoclonal [SERPINB5/4978] antibody to MASPIN for WB and IHC-P with samples derived from human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: MASPIN
Clonality: Monoclonal
Clone: SERPINB5/4978
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR