Anti-ROR gamma Rabbit Recombinant Antibody [clone: RORC/8017R]

Lieferant: ANTIBODIES.COM
A316937-100
ANTIA316937-100EA 680 EUR
ANTIA316937-100
Anti-ROR gamma Rabbit Recombinant Antibody [clone: RORC/8017R]
Antikörper
Recombinant rabbit monoclonal [RORC/8017R] antibody to ROR gamma for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: ROR gamma
Clonality: Recombinant
Clone: RORC/8017R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR