Anti-SPARC Mouse Monoclonal Antibody [clone: OSTN/3756]

Lieferant: ANTIBODIES.COM
A317055-100
ANTIA317055-100EA 680 EUR
ANTIA317055-100
Anti-SPARC Mouse Monoclonal Antibody [clone: OSTN/3756]
Antikörper
Mouse monoclonal [OSTN/3756] antibody to SPARC for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: SPARC
Clonality: Monoclonal
Clone: OSTN/3756
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR