Anti-SPARC Rabbit Recombinant Antibody [clone: OSTN/8528R]

Lieferant: ANTIBODIES.COM
A317057-100
ANTIA317057-100EA 680 EUR
ANTIA317057-100
Anti-SPARC Rabbit Recombinant Antibody [clone: OSTN/8528R]
Antikörper
Recombinant rabbit monoclonal [OSTN/8528R] antibody to SPARC for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: SPARC
Clonality: Recombinant
Clone: OSTN/8528R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR