Anti-IL3RA / CD123 Rabbit Recombinant Antibody [clone: DM31] (Biotin)

Lieferant: ANTIBODIES.COM
A317425-100 A317425-500
ANTIA317425-100EA 925 EUR
ANTIA317425-100 ANTIA317425-500
Anti-IL3RA / CD123 Rabbit Recombinant Antibody [clone: DM31] (Biotin)
Antikörper
Recombinant rabbit monoclonal [DM31] antibody to IL3RA / CD123 (Biotin) for ELISA and Flow Cytometry with samples derived from human.

  • Validated Applications: ELISA, Flow Cytometry
  • Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100

Type: Primary
Antigen: IL3RA / CD123
Clonality: Recombinant
Clone: DM31
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR