Anti-IL3RA/CD123 Rabbit Recombinant Antibody [clone: DM33]

Lieferant: ANTIBODIES.COM
A318661-100 A318661-500
ANTIA318661-100EA 640 EUR
ANTIA318661-100 ANTIA318661-500
Anti-IL3RA/CD123 Rabbit Recombinant Antibody [clone: DM33]
Antikörper
Recombinant rabbit monoclonal [DM33] antibody to IL3RA/CD123 for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, flow cytometry: 1:100

Type: Primary
Antigen: IL3RA/CD123
Clonality: Recombinant
Clone: DM33
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR