Anti-SON Goat Polyclonal Antibody
Lieferant: ANTIBODIES.COM
A82535-100
ANTIA82535-100EA 475 EUR
ANTIA82535-100
Anti-SON Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody to SON for ELISA and IHC with samples derived from human.
  • Validated applications: ELISA, IHC
Type: Primary
Antigen: SON
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human