Anti-CHN2 Goat Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A82874-100
ANTIA82874-100EA 475 EUR
ANTIA82874-100
Anti-CHN2 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody to CHN2 for ELISA and IHC with samples derived from human.

  • Validated applications: ELISA, IHC

Type: Primary
Antigen: CHN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR