Anti-ADH1B Goat Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A83165-100
ANTIA83165-100EA 475 EUR
ANTIA83165-100
Anti-ADH1B Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody to ADH1B for ELISA, WB and IHC with samples derived from human.

  • Validated applications: ELISA, WB, IHC

Type: Primary
Antigen: ADH1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR