Anti-RAD51C Goat Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A83469-100
ANTIA83469-100EA 475 EUR
ANTIA83469-100
Anti-RAD51C Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody to RAD51C for ELISA and WB with samples derived from human.

  • Validated applications: ELISA, WB

Type: Primary
Antigen: RAD51C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR