Anti-UBE2R2 Goat Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A83895-100
ANTIA83895-100EA 475 EUR
ANTIA83895-100
Anti-UBE2R2 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody to UBE2R2 for ELISA, IEM and IHC with samples derived from human and rat.

  • Validated applications: ELISA, IEM, IH

Type: Primary
Antigen: UBE2R2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR