Anti-ADH5 Goat Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A84301-100
ANTIA84301-100EA 475 EUR
ANTIA84301-100
Anti-ADH5 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody to ADH5 for ELISA, WB and IHC with samples derived from human, mouse and rat.

  • Validated applications: ELISA, WB, IH

Type: Primary
Antigen: ADH5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR