Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [Clone: MEM-136]

Lieferant: ANTIBODIES.COM
A85630-100
ANTIA85630-100EA 260 EUR
ANTIA85630-100
Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [Clone: MEM-136]
Antikörper
Mouse monoclonal [MEM-136] antibody to HLA DR + HLA DP for IP, WB, IHC-P and flow cytometry with samples derived from human.

  • Validated applications: IP, WB, IHC-P, Flow cytometry
  • Recommended dilutions: Flow cytometry: 1 to 4 µg/ml

Type: Primary
Antigen: HLA DR + HLA DP
Clonality: Monoclonal
Clone: MEM-136
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR