Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [Clone: HL-38] (FITC (Fluorescein Isothiocyanate))

Lieferant: ANTIBODIES.COM
A85925-100
ANTIA85925-100EA 300 EUR
ANTIA85925-100
Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [Clone: HL-38] (FITC (Fluorescein Isothiocyanate))
Antikörper
Mouse monoclonal [HL-38] antibody to HLA DR + HLA DP (FITC) for flow cytometry with samples derived from human and porcine.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: HLA DR + HLA DP
Clonality: Monoclonal
Clone: HL-38
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Porcine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR